Learn More
Mettler Toledo™ pH/Ion Meter SevenDirect SD50

The SevenDirect SD50 is an accurate and flexible benchtop meter for the determination of pH, ORP and ion concentrations. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.
Brand: Mettler Toledo™ 30671545
2225.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and ion content of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
- Large color touchscreen provides direct measurement and easy data entry.
- Instructions guide users through the sensor calibration process.
- View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
- Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
- The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
- A two-level user management protects the most important settings.
- The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
- Dedicated positions for cleaning and storage support correct sensor handling.

Specifications
0.5 % | |
PC, Printer, USB Stick | |
Single-channel | |
IP54 | |
0.1, 0.01, 0.001 | |
PerfectION Combined Fluoride Electrode | |
Ready-to-measure kit | |
3 or 4 digits (auto-range) | |
1, 0.1 | |
-22°F to +266°F | |
195 x 205 x 65 mm | |
850 g |
0.002 | |
SevenDirect SD50 F-Ion Kit | |
Color TFT | |
-2000 to 2000 mV | |
-30°C to +130°C | |
2000 Standard Measurements | |
-2,000 to 20,000 pX, 0 to 999,999 mg/L (ppm) | |
1, 0.1 mV | |
±0.1°C between 0°C and 100°C | |
0.1°C | |
1.9 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.