Learn More
Mettler Toledo™ pH/Ion Meter SevenDirect SD50
GREENER_CHOICE
The SevenDirect SD50 is an accurate and flexible benchtop meter for the determination of pH, ORP and ion concentrations. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.
£1346.80 - £2310.00
Specifications
Accuracy (Ion) 0.5 %
Accuracy (pH) 0.002
Data Management Details PC, Printer, USB Stick
Channels Single-channel
Display Type Color TFT
View More Specs
Products 7
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17590996
GREENER_CHOICE
View Documents Promotion Details
Mettler Toledo™
30671552
SevenDirect SD50, Meter Only
£1346.80
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
17510996
GREENER_CHOICE
View Documents Promotion Details
Mettler Toledo™
30671544
SevenDirect SD50 Kit
£1585.00
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
17530996
GREENER_CHOICE
View Documents Promotion Details
Mettler Toledo™
30671546
SevenDirect SD50 HA Kit
£1695.00
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
17540996
GREENER_CHOICE
View Documents
Mettler Toledo™
30671547
SevenDirect SD50 Organic
£1815.00
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
17550996
GREENER_CHOICE
View Documents
Mettler Toledo™
30671548
SevenDirect SD50 Pure H2O
£1825.00
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
17560996
GREENER_CHOICE
View Documents Promotion Details
Mettler Toledo™
30671549
SevenDirect SD50 LowVol Kit
£1925.00
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
17520996
GREENER_CHOICE
View Documents
Mettler Toledo™
30671545
SevenDirect SD50 F-Ion Kit
£2310.00
Each
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
Description
Description
SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of pH, ORP and ion content of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.
  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.
Specifications
Specifications
0.5 %
PC, Printer, USB Stick
Color TFT
-2000 to 2000 mV
-30°C to +130°C
-2,000 to 20,000 pX, 0 to 999,999 mg/L (ppm)
±0.1°C between 0°C and 100°C
0.1°C
0.002
Single-channel
IP54
0.1, 0.01, 0.001
2000 Standard Measurements
3 or 4 digits (auto-range)
-22°F to +266°F
195 x 205 x 65 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Mettler Toledo™ pH/Ion Meter SevenDirect SD50