missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Compatible with majority of pH meters
- Electrolyte KCl 4M + AgCl
- Diameter of 6 or 8mm
- Available with 1m BNC cable or DIN connector
- Temp electrode pHast : Recommended for low or high temperature measurements (−5 to +100°C) or during rapid temperature variation
Specifications
DIN | |
-5°C to +100°C | |
6 mm |
0 to 13 | |
115 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Special Offers
- Multiple Manufacturers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ pHast Temp™ Micro-pH Electrode > DIN connector; Dia. x L: 6 x 115mm
Spot an opportunity for improvement?Share a Content Correction