missing translation for 'onlineSavingsMsg'
Learn More
Phenyl p-tolyl sulfone, 98+%, Thermo Scientific Chemicals Product Code.: 11392026

Phenyl p-tolyl sulfone, 98+%, Thermo Scientific Chemicals

Product Code. 11392026
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11392026

Brand: Thermo Scientific Alfa Aesar L03128.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 640-57-3
Molecular Formula C13H12O2S
Molecular Weight (g/mol) 232.297
MDL Number MFCD00025038
InChI Key YBRXHRWLEFCFEG-UHFFFAOYSA-N
Synonym phenyl p-tolyl sulfone, sulfone, phenyl p-tolyl, 4-methylphenyl phenyl sulfone, benzene, 1-methyl-4-phenylsulfonyl, 1-methyl-4-phenylsulfonyl benzene, 4-methyldiphenylsulfone, p-tolyl phenyl sulfone, p-phenylsulphonyl toluene, 1-benzenesulfonyl-4-methylbenzene, 1-benzenesulfonyl-4-methyl-benzeneShow MoreShow Less
PubChem CID 69497
IUPAC Name 1-(benzenesulfonyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 125°C to 129°C
Odor Odorless
Quantity 10 g
Beilstein 1872872
Formula Weight 232.3
Percent Purity ≥98%
Chemical Name or Material Phenyl p-tolyl sulfone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberGBFA030526 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03128.09,L03128.18
Lot Number10116797 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03128.09,L03128.18
Lot Number10213164 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03128.09,L03128.18
Lot Number10234988 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03128.09,L03128.18
Lot Number10204598 Certificate TypeCertificate of Analysis Date02/12/2021 Catalog NumberL03128.09,L03128.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phenyl p-tolyl sulfone, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.