missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 5X), with EDTA

Helps to maintain a constant pH

Brand:  Thermo Scientific Chemicals J60893.K2

58.40 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15433589

  • £58.69 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

PBS with EDTA is also used to disengage attached and clumped cells. PBS-EDTA is used as a buffer in procedures generating dendritic cells from monocytes. PBS+EDTA to get better cell sorting. PBS/EDTA may also be used directly without addition of trypsine for very gentle cell dissociation. PBS can be used to take a reference spectrum when measuring the protein adsorption in ellipsometry. PBS can be used as a diluent in methods to dry biomolecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 X
Phosphate Buffered Saline (PBS)
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM11L575 Certificate TypeCertificate of Analysis Date17/01/2025 Catalog NumberJ60893.K3,J60893.K2
Lot NumberM11L560 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog NumberJ60893.K3,J60893.K2
Lot NumberX23K527 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberJ60893.K3,J60893.K2
Lot NumberQ11K540 Certificate TypeCertificate of Analysis Date24/05/2024 Catalog NumberJ60893.K3,J60893.K2
Lot NumberM16K571 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog NumberJ60893.K3,J60893.K2
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 5X), with EDTA > 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only