missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ pHrodo™ Red and Green Epidermal Growth Factor (EGF) and pHrodo™ Red Transferrin Conjugates

Detect internalized EGF and transferrin in cells with the pHrodo Red and pHrodo Green Epidermal Growth Factor (EGF) and pHrodo Red Transferrin conjugates, for use in applications such as HCS, flow cytometry, microplate-based fluorometry, and HTS.

Brand:  Molecular Probes™ P35376

379.40 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 15362662

  • £542.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Orange
pHrodo™ Red Transferrin Conjugate
Cell Analysis
Red Transferrin Conjugate
Lyophilized Powder
1 mg
560/585 nm
Store at -5° to -30°C, desiccate, and protect from light.
Fluorescence
Fluorescence Microscope, Flow Cytometer, Fluorometer, High Content Analysis Instrument
pHrodo Red
pHrodo™
Endosomes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2978776 Certificate TypeCertificate of Analysis Date22/12/2024 Catalog NumberP35376
Lot Number2785568 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog NumberP35376
Lot Number2725395 Certificate TypeCertificate of Analysis Date17/08/2023 Catalog NumberP35376
Lot Number2616042 Certificate TypeCertificate of Analysis Date05/04/2023 Catalog NumberP35376
Lot Number2566333 Certificate TypeCertificate of Analysis Date29/12/2022 Catalog NumberP35376
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ pHrodo™ Red and Green Epidermal Growth Factor (EGF) and pHrodo™ Red Transferrin Conjugates > Red Transferrin Conjugate

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.