Learn More
Thermo Scientific™ 1.1 mL Microtube System, Racked
Pre-racked 1.1mL-tall microtubes boxed to match a standard 96-well microplate footprint for multichannel pipetting or automated workstations.
Brand: Thermo Scientific™ 15082
Description
Thermo Scientific™ 1.1mL Microtubes and Racked Systems consist of 1.1mL tall microtubes supplied pre-racked in boxes to match a standard 96-well microplate footprint.
The microtubes resembles 1mL pipette tips except that they have round bottoms. Like a box of 1mL pipette tips, the racked microtubes match multi-channel pipettors, providing convenience for a variety of reagent- and sample-dispensing procedures.Highlights:
- 8 x 12 format simplifies and speeds applications in which a multi-channel pipettor is used
- Format is also compatible with robotic workstations
- Thermo Scientific Microtube System can be centrifuged

Specifications
Pierce 1.1mL Microtube Racked System | |
Thermo Scientific™ | |
Microtube Racked System | |
1.1 mL |
Store at room temperature. | |
10 boxes | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZA4114591 | Certificate of Analysis | 02/02/2024 | 15082 |
YJ4017201 | Certificate of Analysis | 01/11/2023 | 15082 |
YH3897212 | Certificate of Analysis | 10/08/2023 | 15082 |
YH3897212A | Certificate of Analysis | 10/08/2023 | 15082 |
XA3480781 | Certificate of Analysis | 30/11/2022 | 15082 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.