Learn More
Thermo Scientific™ Immuno Tubes and Stoppers

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.
Brand: Thermo Scientific™ 444202
180.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Reproduce results time after time with Thermo Scientific™ Immuno Tubes and Stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays. These products are backed by industry-leading quality standards and certifications.
Features:
- Offered in choice of three surfaces for optimal binding
- Three tube sizes with stoppers available (ordered separately)
- Fits standard equipment
Choice of Surface:
- Thermo Scientific™ PolySorp™: a hydrophobic surface that preferentially binds biomolecules that have hydrophobic domains
- Thermo Scientific™ MaxiSorp™: a hydrophilic surface that is optimized for binding IgG and also displays increased binding of many other proteins and biomolecules that possess hydrophilic/hydrophobic character
- Thermo Scientific™ Mini-Sorp™: a polyethylene-based material that provides a low-binding surface
Liquid Phase Immunoassay Tubes (Cat. Nos. 443990, 468608):
- MiniSorp surface
- Molded from special polyethylene formulation
- For liquid phase immunoasssays, including RIA, for storing of reagents or collecting chromatographic fractions
Solid Phase Immunoassay Tubes:
- PolySorp and MaxiSorp surfaces available for choice of optimal binding
- For solid phase immuno techniques such as IRMA, ELISA and ILMA
- Certified binding homogeneity
- High-quality polystyrene for optimal quality
- StarTube design gives higher sensitivity and faster assay times
- Enhances assay sensitivity and reduces incubation times
Recommended for:
- ELISA
- Solid Phase RIA
- Liquid Phase RIA
- Sample storage

Specifications
Polystyrene | |
MaxiSorp | |
12 mm | |
3000 | |
Immuno Tube |
Immuno Tubes and Stopper | |
75 mm | |
100 | |
5 mL |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
183752 | Certificate of Compliance | 06/02/2025 | 444202 |
169829 | Certificate of Compliance | 03/12/2024 | 444202 |
178113 | Certificate of Compliance | 03/12/2024 | 444202 |
183752 | Certificate of Compliance | 03/12/2024 | 444202 |
172655 | Certificate of Compliance | 03/12/2024 | 444202 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.