Learn More
MP Biomedicals™ Cell Lysis Solution for Tissues and Cells
Cell Lysis/DNA solubilizing solution for tissues and cell extractions. Used in conjunction with a Protein Precipitation Solution (PPS). Component of the FastDNA Kit and FastDNA SPIN Kit.
Brand: MP Biomedicals™ 116540409
Description
- CLS-TC is a colorless, precipitate-free solution, with a pH range of 5.8 to 6.2
- This reagent is stable at room temperature
- Shelf life can be extended by storing at 4°C

Specifications
Store at 2°C to 8°C | |
Nucleic Acid Isolation and Purification | |
CLS-TC |
Kit | |
5.8 to 6.2 | |
110 mL |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- CLS-TC
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium dodecyl sulphate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only (RUO). Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.