missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Clear Milk Blocking Buffer (10X)

Thermo Scientific™ Pierce Clear Milk Blocking Buffer is a long shelf-life, high performance replacement for homemade milk blocking buffers in Western blotting applications.

Brand:  Thermo Scientific™ 37587

139.16 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 13494209

  • £203.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Clear Milk Blocking Buffer is a long shelf-life, high performance replacement for homemade milk blocking buffers in Western blotting applications.

Additional Information and Online Ordering

The Thermo Scientific Pierce Clear Milk Blocking Buffer (10X) is a proprietary pre-formulated milk solution for blocking excess nonspecific binding sites, reducing background in Western blotting applications and diluting antibodies when used with nitrocellulose and PVDF membranes. Pierce Clear Milk Blocking Buffer (10X) provides lower background, enhanced sensitivity, extended shelf life and reproducible results compared to homemade buffers based on dissolved non-fat dry milk.

Highlights:

  • Excellent stability – stable for 1 year stored at 4°C, unlike typical homemade milk buffers

  • Convenient – concentrated formulation saves storage space and can be diluted easily to obtain optimal blocking results for specific applications

  • Easy to use – no waiting for powdered milk to dissolve with this ready–to–dilute solution

  • Popular – nonfat milk has been used for many years in a variety of protein methods, although it is not recommended for avidin-based techniques because it contains some endogenous biotin

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blocking Buffer
Liquid
10X
Clarified milk proteins
100 mL
Western Blot
Borate
Upon receipt store at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA4548881 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number37587
Lot NumberZJ4490631 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number37587
Lot NumberZH4296741 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog Number37587
Lot NumberZC4146271 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog Number37587
Lot NumberZB4139391 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number37587
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Clear Milk Blocking Buffer (10X) > 100 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.