missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ GST Agarose

Isolate or remove cross-reacting GST-specific serum or other antibody solutions with immobilized glutathione S-transferase agarose.

Brand:  Thermo Scientific™ 20211

Product Code. 11816734

  • £644.00 / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Perform isolation or removal of cross-reacting GST-specific antibodies from serum or other antibody solutions with Thermo Scientific™ Pierce GST Agarose.

This GST resin consists of purified glutathione S-transferase protein from Schistosoma japonicum that has been immobilized on crosslinked 6% beaded agarose. Antibody molecules in that cross-react with the GST portion of a recombinant fusion protein can be removed from the desired protein-specific antibody by passing antiserum over a GST agarose column. GST antibodies will bind to the immobilized GST, whereas antibodies against other fusion proteins will pass through the column unrestrained.

Highlights:

  • Specific – provides purification of target antibodies from those directed against the GST tag

  • Reusable – bound antibodies can be eluted with 6M guanidinium chloride allowing for 10 purifications with each column or aliquot of resin

  • Two formats – choose either pre-packed columns or resin slurry for use at a variety of scales

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Agarose
Agarose Resin, Affinity
GST Protein
Pierce GST Agarose
5 mL
Pierce™
Bottle
Anti-GST Antibody
4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZD387657 Certificate TypeCertificate of Analysis Date30/04/2024 Catalog Number20211
Lot NumberYI382529 Certificate TypeCertificate of Analysis Date03/10/2023 Catalog Number20211
Lot NumberYG379550 Certificate TypeCertificate of Analysis Date05/08/2023 Catalog Number20211
Lot NumberYB369130 Certificate TypeCertificate of Analysis Date09/03/2023 Catalog Number20211
Lot NumberXK363213 Certificate TypeCertificate of Analysis Date19/11/2022 Catalog Number20211
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ GST Agarose > Resin; 5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.