Learn More
Abeba™ Rubber 5200 Shoes
ESD dissipative shoe is acid and base resistant.
Brand:  Abeba™ 5200/42
 View more versions of this product
Product Code. 15356691
  • £61.15 / 1 pair
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB SRC
  • Smooth leather white
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Specifications
CE EN ISO 20347:2012 OB SRC
Leather
Slip-resistant
5200
White
42
Rubber
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abeba™ Rubber 5200 Shoes > Size: 42