missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Spin Columns - Snap Cap

Perform small-scale affinity methods, such as immunoprecipitation assays with agarose beads using these empty spin columns and cups (5 to 500μL).

Brand:  Thermo Scientific™ 69725

138.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10351454

  • £156.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Pierce Spin Columns - Snap Cap
50 columns
Microcentrifuge, Spin Column
Pierce™
Spin Column
Empty Column, Polyethylene Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL403242 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number69725
Lot NumberZJ392787 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number69725
Lot NumberZJ392786 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog Number69725
Lot NumberZI392785 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number69725
Lot NumberZG392784 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog Number69725
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Spin Columns - Snap Cap > Spin Columns; 1mL capacity

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.