Learn More
Cytiva Microspin Column
For the rapid purification of DNA for use in a wide range of applications
Brand:  Cytiva 27-3565-01
137.49 GBP valid until 2025-12-31
Use promo code "24339" to get your promotional price.
Product Code. 11783299
  • £150.52 / Pack of 100
Explore more special offers
Description
Description
  • Empty microspin columns with screw caps for use with chromatography media of choice such as Sephadex™ for desalting, DNA purification etc
  • Empty microspin columns provide flexibilty with media choice and bed volume allowing process optimization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rapid purification of DNA, desalting/buffer exchange of DNA
Column
100 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Microspin Column > pack of 50