missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ NHS-Activated Magnetic Beads

Immobilize proteins or other ligands for magnetic separation affinity purification with these magnetic particles coated with amine-reactive NHS-ester groups.

£312.00 - £1016.00

Specifications

Concentration Slurry: 10 mg/mL, 1% solids
Content And Storage Upon receipt store at 4°C. Product shipped with an ice pack.
Surface Functionality NHS Ester
Particle Size 1 μm
Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12331893
View Documents
Thermo Scientific™
88826
1 mL
£312.00
1mL
Please to purchase this item. Need a web account? Register with us today!
12196490
View Documents
Thermo Scientific™
88827
5 mL
£1016.00
5mL
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Slurry: 10 mg/mL, 1% solids
NHS Ester
Liquid
Recombinant
Magnetic Affinity Bead
Upon receipt store at 4°C. Product shipped with an ice pack.
1 μm
Pierce™
Amine
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ NHS-Activated Magnetic Beads

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.