missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pipe Banding Tape

Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.

Brand:  Brady™ 275201

 View more versions of this product

Product Code. 12709874

  • £44.05 / 1 roll
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.
Specifications

Specifications

ROHS Environmental Compliance, EU Directive 2002/95/EC, CSA Accepted, UL Recognized,
Pipe Banding Tape
Green
Tape
Pipe Marker
1 Roll
-112°F to +230°F
0.0028 in.
33 m x 25 mm
33 m
25 mm
PIPEBANDINGTAPEGREEN25MMX33M
Permanent Acrylic, Permanent
Satin
Polyester
Pipe and valve marking
1 Ea.
-80°C to +110°C
0.071 mm
108.27 ft.
0.98 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Pipe Banding Tape > Pipe Banding Tape; Dimensions (L x W): 33 m x 25 mm; Color: Green

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.