Learn More
Brady™ Pipe Banding Tape
Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.
£41.30 - £153.00
Specifications
Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, CSA Accepted, UL Recognized, |
---|---|
Description | Pipe Banding Tape |
Adhesive Type | Permanent Acrylic, Permanent |
Finish | Satin |
Format | Tape |
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
12769854
|
Brady™
275233 |
Black | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12729864
|
Brady™
275230 |
Brown | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12799864
|
Brady™
275225 |
Green | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12769864
|
Brady™
275231 |
Gray | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12799854
|
Brady™
275227 |
Blue | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12729874
|
Brady™
275228 |
Orange | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12789874
|
Brady™
275226 |
Red | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12759874
|
Brady™
275232 |
Purple | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12719884
|
Brady™
275229 |
White | 33 m x 100 mm |
£153.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12749884
|
Brady™
275224 |
Yellow | 33 m x 100 mm |
£150.00
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12779854
|
Brady™
275209 |
Black | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12709864
|
Brady™
275203 |
Blue | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12749864
|
Brady™
275206 |
Brown | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12709874
|
Brady™
275201 |
Green | 33 m x 25 mm |
£42.05
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12779864
|
Brady™
275207 |
Gray | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12739874
|
Brady™
275204 |
Orange | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12799874
|
Brady™
275202 |
Red | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12729884
|
Brady™
275205 |
White | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12769874
|
Brady™
275208 |
Purple | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12759884
|
Brady™
275200 |
Yellow | 33 m x 25 mm |
£41.30
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12789854
|
Brady™
275221 |
Black | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12719864
|
Brady™
275215 |
Blue | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12759864
|
Brady™
275218 |
Brown | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12719874
|
Brady™
275213 |
Green | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12789864
|
Brady™
275219 |
Gray | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12709884
|
Brady™
275214 |
Red | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12739884
|
Brady™
275217 |
White | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
12749874
|
Brady™
275216 |
Orange | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12779874
|
Brady™
275220 |
Purple | 33 m x 50 mm |
£68.35
1 roll |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
12769884
|
Brady™
275212 |
Yellow | 33 m x 50 mm |
£67.05
1 roll |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
Description
- Solid color pipe banding tape provides 360 degree visibility to improve safety and operational efficiency.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, CSA Accepted, UL Recognized, | |
Permanent Acrylic, Permanent | |
Tape | |
Pipe Marker | |
1 Roll | |
-112°F to +230°F | |
0.0028 in. | |
108.27 ft. |
Pipe Banding Tape | |
Satin | |
Polyester | |
Pipe and valve marking | |
1 Ea. | |
-80°C to +110°C | |
0.071 mm | |
33 m |