missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ Pipe Connection Pieces

These pipe connection pieces provide connection to the central collection point. Place the collecting container or canister where you want it: the pipe system can be exactly configured to suit your needs!

Brand:  SymLine™ 106430

87.19 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10233090

  • £117.00 / Each
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Wide range of connectors, branches and manifolds allow the pipe system to be connected precisely to the conditions in your laboratory
  • Specifications

    Specifications

    32 mm (f) to 32 mm (m)
    Angled
    PTFE-EL Sealing Ring and Knurled Nut
    Connecting Pipes with 32 mm OD
    32 mm
    Black
    150 mm
    HDPE-EC
    Tube System with SymLine™ FlexTube and FlexTubePro, Connectors, Distributors and Shut-off
    Connection for Pipe System
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    One moment while we fetch your results.
    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    SymLine™ Pipe Connection Pieces > Pipe Connection Pieces; Closure Size: 32 mm (f) to 32 mm (m); Design: Angled; Includes: PTFE-EL Sealing Ring and Knurled Nut; Length (Metric): 150 mm

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.