Learn More
SymLine™ Pipe Connection Pieces
These pipe connection pieces provide connection to the central collection point. Place the collecting container or canister where you want it: the pipe system can be exactly configured to suit your needs!
£103.91 - £246.00
Specifications
Closure Color Black
Liner Material HDPE-EC
For Use With (Application) Connecting Pipes with 32 mm OD
For Use With (Equipment) Tube System with SymLine™ FlexTube and FlexTubePro, Connectors, Distributors and Shut-off
Diameter (Metric) Inner 32 mm
View More Specs
Products 11
Product Code Brand Closure Size Design Includes Length (Metric) Price Quantity & Availability  
Product Code Brand Closure Size Design Includes Length (Metric) Price Quantity & Availability  
17818630
View Documents
SymLine™
106690
32 mm (f) Straight Pipe Extension 600 mm
£103.91
Each
15514515
View Documents
SymLine™
106427
32 mm (f) Straight PTFE-EL Sealing Ring and Knurled Nut 118 mm
£133.00
Each
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
10233090
View Documents
SymLine™
106430
32 mm (f) to 32 mm (m) Angled PTFE-EL Sealing Ring and Knurled Nut 150 mm
£117.00
Each
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
10754432
View Documents
SymLine™
106440
32 mm (f) to 32 mm (m) Angled PTFE-EL Sealing Ring and Knurled Nut 500 mm
£131.00
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
14746901
View Documents
SymLine™
106421
32 mm (f) to 32 mm (m) Curved PTFE-EL Sealing Ring and Knurled Nut 90 mm
£123.00
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
15367439
View Documents
SymLine™
106456
32 mm (f) to GL25 (m) Curved PTFE-EL Sealing Ring and Knurled Nut 90 mm
£145.00
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
15813432
View Documents
SymLine™
106458
2 x 32 mm (f), 1 x GL25 (m) T-piece PTFE-EL Sealing Ring and Knurled Nut 118 mm
£239.00
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
13528840
View Documents
SymLine™
106426
3 x 32 mm (f) T-piece PTFE-EL Sealing Ring and Knurled Nut 118 mm
£227.00
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
15844435
View Documents
SymLine™
106476
3 x GL25 (m) Manifold Manifold with 2 Fixation Holes 68.25 mm
£142.00
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
15803432
View Documents
SymLine™
106457
32 mm (f) to GL25 (m) Elbow PTFE-EL Sealing Ring and Knurled Nut 75.5 mm
£170.00
Each
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
18686478
View Documents
SymLine™
106732
32 mm (f) Straight Pipe Extension 1200 mm
£246.00
1 sheet
Description
Description
  • Wide range of connectors, branches and manifolds allow the pipe system to be connected precisely to the conditions in your laboratory
  • Specifications
    Specifications
    Black
    Connecting Pipes with 32 mm OD
    32 mm
    HDPE-EC
    Tube System with SymLine™ FlexTube and FlexTubePro, Connectors, Distributors and Shut-off
    Connection for Pipe System