missing translation for 'onlineSavingsMsg'
Learn More
Learn More
piperidine-1-carboximidamide hydroiodide, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 255.1 |
Percent Purity | 97% |
Chemical Name or Material | Piperidine-1-carboximidamide hydroiodide |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
piperidine-1-carboximidamide hydroiodide, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction