Learn More
Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces
Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.
£118.76 - £775.00
Product Code | Brand | Product Type | Color | Surface Treatment | Total Volume (Metric) Well | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Color | Surface Treatment | Total Volume (Metric) Well | Price | Quantity & Availability | ||
10143632
|
Thermo Scientific™
436014 |
Microplate | Clear | Streptavidin | 400 μL |
£606.00
Pack of 15 |
In Stock
Log in to see stock. |
||
10387522
|
Thermo Scientific™
436015 |
Microplate | White | Streptavidin | 400 μL |
£540.00
Pack of 15 |
In Stock
Log in to see stock. |
||
10541493
|
Thermo Scientific™
436016 |
Microplate | Black | Streptavidin | 400 μL |
£540.00
Pack of 15 |
In Stock
Log in to see stock. |
||
10133682
|
Thermo Scientific™
436017 |
Microplate | Clear | Streptavidin | 120 μL |
£775.00
Pack of 15 |
In Stock
Log in to see stock. |
||
10788944
|
Thermo Scientific™
436020 |
Solid module F8 (x12) in frame | Clear | Streptavidin | 400 μL |
£625.00
Pack of 15 |
In Stock
Log in to see stock. |
||
10636083
|
Thermo Scientific™
436022 |
Lock-Well module C8 (x12) in frame | Clear | Streptavidin | 350 μL |
£625.00
Pack of 15 |
In Stock
Log in to see stock. |
||
10551493
|
Thermo Scientific™
436018 |
Microplate | White | Streptavidin | 120 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10709144
|
Thermo Scientific™
436032 |
Microplate | Clear | Glutathione | 400 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10723686
|
Thermo Scientific™
95029263 |
Microplate | Clear | Streptavidin | 400 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10604617
|
Thermo Scientific™
95029293 |
Breakable module F8 (x12) in frame | Clear | Streptavidin | 400 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.
Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind):
- Highly specific binding surfaces reduce variability in molecular orientation.
- Immobilizer surfaces improve signal to noise ratios.
- Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules.
Specifications
Unspecified |