Documents
Scientific Resources
Application Notes
- 1
- 2
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Brand: Thermo Scientific™ 436008
622.99 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Standard | |
8 x 12 | |
Covalent | |
Microplate | |
400 μL | |
100 μL | |
Amine | |
Round, Flat Bottom (F-well) | |
5 | |
Peptides |
Fluorescence | |
30 Cases | |
Black | |
Polystyrene | |
96 | |
Non-sterile | |
400 μL | |
30 | |
Unspecified |
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
184898 | Certificate of Compliance | 15/01/2025 | 436008 |
184899 | Certificate of Compliance | 09/01/2025 | 436008 |
178274 | Certificate of Compliance | 03/12/2024 | 436008 |
167766 | Certificate of Compliance | 03/12/2024 | 436008 |
164461 | Certificate of Compliance | 03/12/2024 | 436008 |
Expires: 2025-12-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.