missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Polycarbonate Face Shield Product Code.: 13242963
Add to basket

Thermo Scientific™ Polycarbonate Face Shield
GREENER_CHOICE

Product Code. 13242963
Clear, 11.6 in., 7.8 in.
Click to view available options
Lens Tint:
Clear
Length (English):
11.6 in.
Width (English):
7.8 in.

Product Code. 13242963

Brand: Thermo Scientific™ 63550001

Edge
Add to basket
Add to basket

Wear this adjustable polycarbonate face shield over safety glasses. It provides impact protection and features an optically clear 1/16 in. thick polycarbonate window.

Wear the adjustable Thermo Scientific™ Nalgene™ Polycarbonate Faceshield over safety glasses. With an optically clear window made of 1/16 in. (1.5mm) thick polycarbonate, this faceshield provides impact protection as well as protection from UV light in the 200-360nm range. It is useful in germicidal procedures as well as reading of chromatographic/electrophoretic strips and gels. Complies with OSHA 29CFR1910.1030 and ANSI Z87.1-1989 standards for use with bloodborne pathogens.

Features:
  • Wraparound design offers both front and side protection while height provides coverage from the bottom of the neck to the top of the head
  • Features a comfortable and adjustable polyethylene headdress
  • Biohazard/Transparent
Note:UV protection from 200 to 360nm (UV radiation may cause the polycarbonate to develop a slight yellow/brown tint.)
Note: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 

 

 

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Thickness (English) 0.060 in.
Thickness (Metric) 1.5 mm
Width (English) 7.8 in.
Description Adjustable Face Shield
Lens Tint Clear
Product Type Shield Face PC/PE
Length (English) 11.6 in.
No. per Case 4/Cs.
Material Polycarbonate
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now

Note: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Polycarbonate Face Shield >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.