Learn More
Buerkle™ Polypropylene Compact Spigot
Saves space with compact design. Buerkle™ Polypropylene Compact Spigot for canisters and barrels is ideal for use within refrigerators and on racks.
Brand:  Buerkle™ 0530-1000
28.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11911461
  • £31.05 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
A compact spigot for canisters and barrels ideal for use within refrigerators and on racks, etc. The short discharge does not protrude over the lower edge of the container, making it the perfect choice especially when space is very limited.
  • Space-saving design
  • Optimised discharge for even flow
  • With connecting union nut for vertical position of the stop cock
  • Tight-closing for complete seal
Specifications
Specifications
Compact Spigot
Polypropylene
19.05 mm/GL14
Canisters, Barrels, Ideal Addition to Flachmann Jerry Cans
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Polypropylene Compact Spigot > Inner Thread Size: 3/4in.