Learn More
Masterflex™ Polypropylene ADCF Luer Fittings
Ensure process integrity with full material traceability. Cole-Parmer™ Polypropylene ADCF Luer Fittings are ideal for a variety of critical applications, including syringes or small tubing. These fittings are inspected to be free from contaminants, and are fully traceable.
Brand: Masterflex™ 30800-30
Description
Ensure process integrity with full material traceability. Cole-ParmerTM Polypropylene ADCF Luer Fittings are ideal for a variety of critical applications, including syringes or small tubing. These fittings are inspected to be free from contaminants, and are fully traceable.
All parts meet tight dimensional tolerances for increased reliability and process purity. Choose from a range of sizes, 1⁄16" to 1⁄4", with a variety of connection options.- Manufactured from 100% virgin raw materials in a Class 100,000 environment
- In compliance with ISO 594-1 standards
- Manufactured of animal-free natural polypropylene
- Meets USP Class VI specifications
- Very good chemical resistance
- Max temperature 190°F
Specifications
Polypropylene | |
Ideal for a variety of critical applications, including syringes or small tubing. | |
Polypropylene ADCF Luer Fitting |
Male luer lock plug | |
25 Pack |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.