missing translation for 'onlineSavingsMsg'
Learn More
Potassium oxalate monohydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10713021

Potassium oxalate monohydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10713021
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10713021

Brand: Thermo Scientific Acros 207715000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrateShow MoreShow Less
PubChem CID 2724193
IUPAC Name dipotassium;oxalate;hydrate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 356.0°C
Color White
pH 7 to 8.5 (5% soln. at 20°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula KO2CCO2K·H2O
Quantity 500 g
Merck Index 15, 7772
Solubility Information Solubility in water: 364g/L (20°C).
Formula Weight 184.24
Percent Purity 99+%
Grade Analysis
Physical Form Crystals or Flakes
Chemical Name or Material Potassium oxalate monohydrate, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468234 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number207710025,207715000
Lot NumberA0468235 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number207710025,207715000
Lot NumberA0466626 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number207710025,207715000
Lot NumberA0462784 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number207710025,207715000
Lot NumberA0454837 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number207710025,207715000
compliance-icons
Product Identifier
  • Potassium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium oxalate monohydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.