missing translation for 'onlineSavingsMsg'
Learn More
Potassium phosphate, monobasic, 99+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10035810

Potassium phosphate, monobasic, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10035810
500 g, Plastic bottle
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10035810

Brand: Thermo Scientific Acros 424205000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphateShow MoreShow Less
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253.0°C
Color Colorless to White
pH 4.1 to 4.5 (5% soln. at 25°C)
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99+%
Linear Formula KH2PO4
Merck Index 15, 7780
Solubility Information Solubility in water: 222g/L (20°C). Other solubilities: insoluble in alcohol
IUPAC Name potassium dihydrogen phosphate
Formula Weight 136.09
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 0.2% max. (2 g, 105°C, 2 hrs)
Packaging Plastic bottle
Decomposition Information >250°C
Chemical Name or Material Potassium phosphate, monobasic
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465550 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number424200250,424200025,424205000
Lot NumberB0155103 Certificate TypeCertificate of Analysis Date09/07/2024 Catalog Number424200250,424200025,424205000
Lot NumberA0457361 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number424200250,424200025,424205000
Lot NumberA0457362 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number424200250,424200025,424205000
Lot NumberB0155523 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number424200250,424200025,424205000
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    14

    Hours

    3

    Minutes

    37

    Seconds

    0
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium phosphate, monobasic, 99+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.