Learn More
OHAUS™ Precision Balance - PR Series
Designed for Routine Weighing Applications in Your Workplace
Brand:  OHAUS™ 30524386
872.17 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 16660324
  • £1180.00 / Each
Explore more special offers
Description
Description
  • The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage, and internal calibration (in select models), making it ideal for routine weighing applications.
  • Equipped with an easy-to-read, bright backlit display and a simple user interface, the PR is incredibly easy to operate, with almost no training required.
  • The PR's small footprint saves desktop space while providing a large weighing surface. The PR is durably constructed, and features a stainless steel pan to withstand day-to-day use in the workplace.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
OHAUS™ Precision Balance - PR Series > Model: PR223M