missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Probenecid, Water Soluble

Probenecid, Water Soluble

Brand:  Molecular Probes™ P36400

235.60 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.



Product Code. 11529306

  • £314.00 / 770mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Probenecid is commonly used to inhibit organic-anion transporters located in the cell membrane. Such transporters can extrude dyes and indicators and thus contribute to poor loading or a high background signal in assays based on retention of the dyes or indicators inside cells. The use of probenecid to block the efflux of intracellular dyes was first described by Di Virgilio et al. (1990), and it has been used with a wide range of anionic dyes and conjugates. The commonly used free acid form of probenecid is difficult to dissolve, requiring 1 M NaOH to get it into solution. Our water-soluble probenecid (P36400) dissolves quickly in assay buffer and eliminates the need to handle caustic NaOH. The water-soluble probenecid is included in the Fluo-4 NW Calcium Assay Kits (F36205, F36206) and is also available separately.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 x 77 mg
Room Temperature
Solid
Cell Labeling Reagent
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2935434 Certificate TypeCertificate of Analysis Date06/07/2024 Catalog NumberP36400
Lot Number2810926 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberP36400
Lot Number2743073 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog NumberP36400
Lot Number2600149 Certificate TypeCertificate of Analysis Date31/05/2023 Catalog NumberP36400
Lot Number2549291 Certificate TypeCertificate of Analysis Date04/11/2022 Catalog NumberP36400
Special Offers

Special Offers

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    28

    Hours

    1

    Minutes

    40

    Seconds

    16
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Probenecid, Water Soluble > 10 x 77mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.