Learn More
Contec™ PROSAT™ Presaturated Nonwoven Wipes

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools with PROSAT™ Presaturated Nonwoven Wipes.
Brand: Contec™ PS-911ETOH
446.99 GBP valid until 2025-12-31
Use promo code "24309" to get your promotional price.
Description
- Consists of 100% polypropylene wipes, saturated with solution of 70% denatured ethanol and 30% deionized water in resealable pouch
- Clean, being free from additives of any kind, and has very low concentration of sodium
- Increase solvent control and accountability and reduce VOC emissions
- Compatible with ISO Class 5 cleanrooms

Specifications
Pre-saturated Wipe | |
23 cm | |
28 cm | |
Not Autoclavable | |
Single-use | |
1-ply | |
Non-sterile |
9 in. | |
11 in. | |
Polypropylene | |
Meltblown Nonwoven | |
23 x 28 cm (9 x 11 in.) | |
70% Denatured Ethanol/ 30% DI Water |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- CONTEC PROSAT MBPP WITH 70% DENATURED ETHANOL
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry extingushing powder to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P411-Store at temperatures not exceeding 40°C/°F.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, methanol, propan-2-ol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1089634-0-0-1 | Certificate of Quality | 10/01/2025 | PS-911ETOH |
1028996-0-0-1 | Certificate of Quality | 26/09/2024 | PS-911ETOH |
992541-0-0-1 | Certificate of Quality | 25/05/2024 | PS-911ETOH |
971366-0-0-1 | Certificate of Quality | 04/11/2023 | PS-911ETOH |
956461-0-0-1 | Certificate of Quality | 10/03/2023 | PS-911ETOH |
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Days
36Hours
14Minutes
46Seconds
42New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.