Learn More
PURUS™ Frames for Biohybrid™ Contamination Control Mats
Designed to be used with the PURUS™ EnviroTack™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.
Brand:  PURUS™ PF 3862 01 W 00
Product Code. 15392002
  • £336.00 / Each
Estimated Shipment: 10-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easily moved for cleaning
  • Non-skid backing ensures frame stays in place
  • Fits all PURUS&trade Biohybrid&trade mats
  • Strong and Durable
  • Non-Slip made from Polymer Coated polyester textile
  • Naturally antimicrobial, mold and mildew resistant
Specifications
Specifications
965 x 1575 mm
914 x 1524 mm PURUS™ Biohybrid™ Mats
White
For use in Pharmaceutical, Biotech, Semiconductor, Cleanrooms, and Industrial Applications
3.175 mm
Polystyrene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
PURUS™ Frames for Biohybrid™ Contamination Control Mats > 965 x 1575 mm