Learn More
Cole-Parmer PVC Lab and Vacuum Tubing
Smooth, polished inner wall prevents buildups—ideal for general laboratory applications
Brand:  Cole-Parmer 50121-20
 View more versions of this product
Product Code. 30053748
  • £985.00 / 15m
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Versatile applications including peristaltic pumps, analytical instruments, incubators, gas, and drain lines. Glass-smooth inner bore design for easy clean-ups and changeovers, enhancing workflow efficiency. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Specifications
3/8 in.
7/8 in.
Translucent
PVC
50 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer PVC Lab and Vacuum Tubing > 3/8 in., 7/8 in.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.