Description
- Complies with ISO 4796
- Borosilicate glass with unground neck
- For use with rubber stoppers (not supplied)
Specifications
Specifications
Capacity (Metric) | 10 L |
Material | Borosilicate Glass |
Certifications/Compliance | ISO 4796 |
Mouth | Unground Wide |
Product Type | Reagent Bottle |
For Use With (Equipment) | Rubber Stoppers |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Product Title
Pyrex™ Wide Neck Reagent Bottle >
Spot an opportunity for improvement?Share a Content Correction