missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Beta Block for 4 x 1.5ml Eppendorf tubes (5 x 3.5 x 14cm)
- Safety containers/blocks for protection against Beta radiation
- Covers are also available
- For holding and protecting 4x 1.5mL acrylic microcentrifuge tubes
Specifications
5 x 3.5 x 14cm | |
For 4 x 1.5ml Eppendorf tubes (5 x 3.5 x 14cm) |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Radiation Safety Blocks Beta > Dimensions: 5 x 3.5 x 14cm; Capacity: 4x 1.5mL
Spot an opportunity for improvement?Share a Content Correction