missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MarqMetrix All-In-One Process Raman Analyzer

The Thermo Scientific™ MarqMetrix™ All-In-One Process Raman Analyzer is a compact and portable Raman spectrometer all-in-one system with patented technology, purpose-built for rapid deployment, ease-of-use, and flexible scalability in markets where time-to-results is key.

Brand:  Thermo Scientific™ 800-04991-01

 View more versions of this product

Product Code. 17719417

  • £73085.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

25 cm
Input voltage: 90-264 VAC, 50-60 Hz, Operating voltage: 12VDC, 2A
6.5 cm-1 average across spectral range
100 to 3250 cm-1
25 cm
Thermally stabilized CCD (2 stage TEC)
Wireless mouse, HDMI cable, monitor
≤16.5 counts at 10 sec. integration
MarqMetrix Process Analyzer with Process Ball Probe
7.7 cm
Software: MarqMetrix Raman suiteController: Integrated PC (Windows 10 OS)
785 nm, Excitation power: 10 to 450 mW
1 keyed Laser interlock switch, Electronic laser trigger
FC/PC (Both excitation and collection)
Laser Certification: Class 3B laser deviceReported to the FDA
1RJ-45 Ethernet port 3 USB 3.0, 1 Display port 1.1 (VGA, DVI or HDMI with adaptor)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MarqMetrix All-In-One Process Raman Analyzer > MarqMetrix All-In-One Process Raman Analyzer with Process Ball Probe

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.