missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Rapid SARS-CoV-2 Antigen Test

The Rapid SARS-CoV-2 Antigen Test Card is an in-vitro diagnostic test for private use. It is used to detect SARS-CoV-2 antigens from anterior nasal swabs.

Brand:  MP Biomedicals 07AG6001BS-DE-EN-FR

Product Code. 17287736

  • £4.19 / 1 test

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Easy sampling using anterior nasal swab specimens
  • Results in 15 minutes
  • Works with a nasal swab of 2.5 cm
  • Specificity 99.20%, Sensitivity 96.77%, Accuracy 98.72
  • Includes all required consumable materials
  • No cross-contamination with other coronaviruses like MERS, 229E, NL63, OC43 or influenza A, or B

Usage Statement: Test for self-testing by laypersons with CE certification

Specifications

Specifications

CE certified self-test for private use/home use
SARS-CoV-2 Nucleocapsid Protein (N protein)
0.9677
Rapid Qualitative Determination of SARS-Cov-2 Virus Antigen
1 Test
Results in 15 minutes
4°C to 30°C
Nasal Swab
Rapid Test
SARS-CoV-2 Antigen Test Cassette (sealed foil pouch), Sterile Swab, Extraction Tube, Extraction Solution, Instructions for Use
0.992
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals Rapid SARS-CoV-2 Antigen Test > 1 Test

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.