missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12255-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5′-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5′ UTR, but encoding the same protein, have been identified for this gene.Specifications
RCAS1 | |
Polyclonal | |
Unconjugated | |
EBAG9 | |
EB9, EBAG9, PDAF, RCAS1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
299864, 55960, 9166 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00559, Q5PQP2, Q9D0V7 | |
Ebag9 | |
RCAS1 Fusion Protein Ag2905 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RCAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction