missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(XII) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XII) chain, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_004361.3 |
---|---|
Gene ID (Entrez) | 1303 |
Name | Collagen alpha-1(XII) chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16062211
|
enQuireBio™
QP8023-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082211
|
enQuireBio™
QP8023-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16072211
|
enQuireBio™
QP8023-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092211
|
enQuireBio™
QP8023-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002221
|
enQuireBio™
QP8023-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012221
|
enQuireBio™
QP8023-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_004361.3 | |
Collagen alpha-1(XII) chain Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1303 | |
Research Use Only | |
COL12A1 | |
Recombinant Protein | |
E. coli | |
DLVFLVDGSW SVGRNNFKYI LDFIAALVSA FDIGEEKTRV GVVQYSSDTR TEFNLNQYYQ RDELLAAIKK IPYKGGNTMT GDAIDYLVKN TFTESAGARV GFPKVAIIIT DGKSQDEVEI PARELRNVGV EVFSLGIKAA DAKELKQIAS TPSLNHVFNV ANFDAIVDIQ NEIISQV | |
Greater than 90% as determined by SDS-PAGE. |