Learn More
Thermo Scientific™ PathoDx™ Respiratory Virus Panel Control Slide
Use PathoDx™ Respiratory Virus Panel Control Slide along with RVP kit to perform direct immunofluorescence test.
Brand: Thermo Scientific™ R62414
Description
Use Thermo Scientific™ Remel™ PathoDx™ Respiratory Virus Panel Control Slide along with PathoDx Respiratory Virus Panel (RVP) kit while performing direct immunofluorescence tests. Viral respiratory diseases in children and adults cause significant morbidity and mortality, especially in young children and older adults. Respiratory specimens (nasopharyngeal aspirates or swabs) are tested for the presence of viral antigen. Culture isolation/confirmation is the standard method for most clinical virology laboratories.
The Negative Control Reagent contains fluorescein-labeled murine antibodies which do not react with the viral agents.
- Ready to use - Prepared reagents
- Easy to store - Store at 2-8°C
- Easy to interpret - Uninfected cells will stain red due to the Evans blue counterstain in each staining reagent
Negative Control Reagent (Red Cap) is one dropper bottle containing 10.0 mL of fluorescein-labeled purified murine IgG antibodies that are non-reactive with any respiratory virus, and Evans blue counterstain in a protein-stabilized buffer solution, with 0.098% sodium azide as preservative.
Each control slide has eight wells and each well is clearly marked with the type of virus-infected cell that is on the slide. One of the wells contains uninfected LLC-MK2 cells and the slide is fixed in acetone.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
PathoDx™ Respiratory Virus Panel Control Slides | |
Control | |
5 Slides/Pk. |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.