Learn More
Thermo Scientific™ Remel™ PathoDxtra™ Strep Grouping Kit
Eliminates need for incubation and provides a faster strep grouping solution than more traditional enzyme extraction.
Brand: Thermo Scientific™ DR0705G
46.96 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Eliminate the need for incubation and provide a faster strep grouping solution than more traditional enzyme extraction with the Thermo Scientific™ PathoDxtra™ Strep Grouping Kit.
- Improves workflow efficiencies — Instant acid extraction without the need for incubation or reconstitution
- Saves time — Simple, rapid procedure produces easy-to-read results
- Reduces costs — Identifies all clinically important Strep groups, including Group D; no additional reagents required
- Provides flexibility — Pack variants available to suit testing needs
Includes:
PathoDxtra Strep Grouping Kit (Mfr. No. DR0700M): 3mL each of A, B, C, D, F and G grouping latex; Polyvalent Positive Control, 2.8mL; Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; Mixing Sticks; Disposable Reaction Cards
PathoDxtra Extraction Reagent Set (Mfr. No. DR0709M): Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; For use with the PathoDxtra Strep Grouping Kit
PathoDxtra Strep Grouping Reagent Set (Mfr. No. DR0710M): Extraction Reagent 1 - 4mL; Extraction Reagent 2 - 4mL; Extraction Reagent 3 - 2 x 10mL; Mixing Sticks, Disposable Reaction Cards; For use with selected, separately purchased PathoDxtra Strep Grouping latex reagents

Specifications
Positive | |
beta-haemolytic streptococci | |
Enzyme Immunoassay Detection | |
Group A, B, C, D, F and G |
PathoDxtra Strep Group F Latex | |
Streptococcus Test Kit | |
Polyvalent | |
60 Tests |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3813467 | Certificate of Analysis | 05/11/2024 | DR0705G |
3778940 | Certificate of Analysis | 21/05/2024 | DR0705G |
3766590 | Certificate of Analysis | 21/03/2024 | DR0705G |
3672338 | Certificate of Analysis | 14/10/2023 | DR0705G |
3615815 | Certificate of Analysis | 14/09/2023 | DR0705G |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Rapid Streptococcal grouping tests correlate best with reference methods when only Streptococci that are Β-hemolytic on sheep blood agar are tested. 1, 2, 3, 4