Learn More
Fisherbrand™ Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector
Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Brand:  Fisherbrand™ 11941-12
488.71 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.
 View more versions of this product
Product Code. 15192738
  • £565.00 / Each
Explore more special offers
Description
Description
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Compliance
WEEE and ROHS compliant
Specifications
Specifications
Deuterium
0.16 kg
Agilent 1100 DAD, 8453 UV and Visible HPLC Detector
0.36 lb.
Item 14-500-721 is comparable to Agilent part number 2140-0605.
Detector Lamp
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Replacement Deuterium Lamp for Agilent UV and Visible HPLC Detector > Deuterium Lamp; For use with: Agilent 1100 DAD, 8453 UV and Visible HPLC Detector