Learn More
Invitrogen™ RiboMinus™ Eukaryote Kit v2
The RiboMinus™ Eukaryote Kit v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA.
Brand: Invitrogen™ A15020
891.10 GBP valid until 2025-12-31
Use promo code "24340" to get your promotional price.
Description
Features of the kit include:
• Seamless integration with RiboMinus™Concentration Module (available separately) to purify RNA after ribosomal RNA depletion
• 1.5-hour workflow
• Input range of 1-5 μg
The RiboMinus™ Eukaryote Kit v2 improves upon the previous system (RiboMinus™ Eukaryote Kit for RNA-Seq) by removing significantly more rRNA (both cytoplasmic and mitochondrial) from the total RNA. When used with our library construction kits for sequencing, the RiboMinus™ Eukaryote Kit v2 provides maximum mapping to reference database with minimum bias, maintaining relative gene expression levels of even smaller RNA species. The resulting total RNA after depletion contains minimum contaminating rRNA with reduced variability.
The RiboMinus Eukaryote Kit v2 is designed for the removal of human, mouse, and rat rRNA. The RiboMinus™ Concentration Module (available separately) is used for RNA purification after rRNA depletion. For magnetic bead-based purification, please see the RiboMinus™ Eukaryote System v2. The RiboMinus Eukaryote Kit v2 has an RNA input range of 1-5 μg. For lower inputs (down to 100 ng), the Low Input RiboMinus™ Eukaryote System v2 is recommended.
The RiboMinus™ Eukaryote Kit v2 contains sufficient reagents to perform 12 ribosomal RNA depletions.
Specifications
• RiboMinus™ Eukaryote Probe Mix, 2-20°C |
|
Human, Mouse, Rat | |
Ion PGM™ System, Ion Proton™ System, Illumina™ Genome Analyzer II (GA II) | |
12 reactions | |
1 - 5 μg total RNA |
Magnetic Bead | |
Transcriptome RNA | |
RiboMinus™ | |
Transcriptome Sequencing | |
Sample Enrichment |
Product Suggestions
Customers who viewed this item also viewed