missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ringer's Solution, lactate-buffered

Brand:  Thermo Scientific Chemicals J67572.K2

152.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15408769

  • £157.34 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ringer's Solution
1 L
Plastic bottle
Liquid
dihydrogen oxide, dihydrogen monoxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY27K516 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog NumberJ67572.AP,J67572.K2
Lot NumberW21K518 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberJ67572.AP,J67572.K2
Lot NumberQ04K545 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog NumberJ67572.AP,J67572.K2
Lot NumberN28K576 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog NumberJ67572.AP,J67572.K2
Lot NumberZ26J587 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberJ67572.AP,J67572.K2
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Ringer's Solution, lactate-buffered > 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only