missing translation for 'onlineSavingsMsg'
Learn More

Bellingham + Stanley™ Rubber Washers for Polarimeter Tubes

Compatible with glass polarimeter tubes. Bellingham + Stanley™ Rubber Washers for Polarimeter Tubes are replacement parts that are placed between the cover glass and end cap of filling tubes used in polarimeters.

Brand:  Bellingham + Stanley™ 35-64

Product Code. 11940088

  • £30.65 / Pack of 12
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Replacement part for glass polarimeter tubes
  • Use between cover glass and end cap
  • 15.5mm diameter rubber washers
  • Sold in packs of 12
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Replacement Washers (used between cover glass and end cap)
Rubber
Glass Polarimeter Tube
15.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bellingham + Stanley™ Rubber Washers for Polarimeter Tubes > Washer diameter: 15.5mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.