Learn More
Thermo Scientific™ S140-AT Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
Support a variety of applications with this rotor, which operates at a maximum of 1,048,000 x g.
Brand: Thermo Scientific™ 45978
16358.53 GBP valid until 2025-06-30
Use promo code "24069" to get your promotional price.
Description
Support applications that require extreme force with the Thermo Scientific™ S140-AT Fixed Angle Rotor; this rotor is capable of operating at over 1,048,000 x g.
Features:
- Fixed angle (35°) titanium rotor
- Capacity: 10 × 2mL
- Maximum Speed/RCF: 140,000rpm/1,048,000 x g
- K factor: 5
Compatible with:
Thermo Scientific™ Sorvall™ MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge
Recommended for:
For rapid separation of serum lipoproteins, DNA, RNA and subcellular fractionation.

Specifications
S140-AT Fixed Angle Rotor | |
Fixed Angle Rotor | |
1,048,684 x g | |
47.9 mm | |
5 | |
11 mm | |
Thermo Scientific™ Sorvall™ MTX and MX Plus micro-ultraCentrifuges | |
713,718 x g |
Titanium | |
10 x 2 mL | |
140,000 rpm | |
32.6 mm | |
34 mm | |
1.8 lb. (0.8 kg) | |
35° |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Exclusive Promotion on Equipment and Instruments
Take advantage of our limited-time offer on top-quality equipment and instruments to elevate your research and laboratory capabilities.Promo Code: 24069Expires: 2025-06-30
- Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.