missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System
Safe and approved storage of flammable hazardous substances in working areas.
POA
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Color | Light Gray |
Material | Steel |
Product Type | Safety storage cabinet |
For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
Alert:
For indoor use only
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Price | Quantity & Availability | |||||
15962846
|
asecos™
3012200133753 |
2 x drawer (sheet steel powder-coated) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15982836
|
asecos™
3012200133744 |
2 x shelf, 1 x perforated insert (285 mm), 1 x bottom collecting sump (sheet steel powder-coated) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15972846
|
asecos™
3012200133754 |
3 Drawers |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15904346
|
asecos™
3034800133848 |
3 Shelves, 1 Perforated Insert, 1 Bottom Collecting Sump, 3 Tray Shelves, 1 Bottom Collecting Sump |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15104609
|
asecos™
3011600130127 |
3 Shelves, 1 Perforated Insert (375 mm), 1 x 33 L Bottom Collecting Sump |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15982536
|
asecos™
3011600133715 |
3 Shelves, 1 Perforated Insert (380 mm), 1 33 L Bottom Collecting Sump |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15912546
|
asecos™
3011600133719 |
3 Tray Shelves, 1 Perforated Insert (365 mm), 1 55.5 L Bottom Collecting Sump |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947466
|
asecos™
3253200133695 |
4 x 6 L Drawers |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15983006
|
asecos™
3020500133709 |
4 x 19 liter drawers (sheet steel powder-coated) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962546
|
asecos™
3011600133724 |
4 x 29 L Drawers |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15914346
|
asecos™
3034800133849 |
6 x drawer (sheet steel powder-coated), 3 x tray shelf, 1 x bottom collecting sump (PP) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967466
|
asecos™
3253200133697 |
6 x 6 liter drawers (sheet steel powder-coated) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15903016
|
asecos™
3020500133711 |
6 x 19 liter drawers (sheet steel powder-coated) |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15982546
|
asecos™
3011600133726 |
6 x 29 L Drawers |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
Steel | |
Safe and approved storage of flammable hazardous substances in working areas |
Light Gray | |
Safety storage cabinet | |
Powder-coated plain |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System