Learn More
3M™ Safety Glasses 1200E Series
Click to view available options
Frame Color: Gray
Description
- Safety glasses with a lightweight half-frame design
- Adjustable soft padded nose bridge ensures a comfortable fit
- Anti-scratch and anti-fog coating provides increased durability and improved vision
- Supplied with a microfibre pouch
- Grey coloured dark shade lens protects against sun glare
- Lens marking: 5-3.1 3M™ 1 FT
- Frame marking: 3M™ 1200E EN166 FT CE
Specifications
Specifications
Lens Coating | Scratch-resistant |
Lens Tint | Gray |
Includes | Microfiber Pouch |
Model | 1200E Series |
Frame Type | Half frame |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Product Title
3M™ Safety Glasses 1200E Series >
Spot an opportunity for improvement?Share a Content Correction