missing translation for 'onlineSavingsMsg'
Learn More
Sand, pure, 40-100 mesh, Thermo Scientific Chemicals Product Code.: 10692423

Sand, pure, 40-100 mesh, Thermo Scientific Chemicals

Product Code. 10692423
1 kg, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
This item is not returnable. View return policy

Product Code. 10692423

Brand: Thermo Scientific Acros 370940010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Boiling Point 2230.0°C
Melting Point 1610.0°C
Color Beige to Gray
Physical Form Crystals
Quantity 1 kg
Linear Formula SiO2
Merck Index 13,8567
Solubility Information Solubility in water: insoluble.
IUPAC Name silanedione
Formula Weight 60.08
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Sand, 40 to 100 Mesh
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464720 Certificate TypeCertificate of Analysis Date31/07/2024 Catalog Number370940250,370940010,370940050,370942500
Lot NumberA0426754 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number370940250,370940010,370940050,370942500
Lot NumberA0460096 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number370940250,370940010,370940050,370942500
Lot NumberA0455771 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number370940250,370940010,370940050,370942500
Lot NumberA0450448 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number370940250,370940010,370940050,370942500
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    15

    Minutes

    4

    Seconds

    26

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sand, pure, 40-100 mesh, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.