missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ Turbo 15 PES Centrifugal Concentrators
BIOPHARMA

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

£172.00 - £818.00

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
Sample Volume 15 mL
View More Specs

Products 14
Product Code Brand MWCO Quantity Price Quantity & Availability  
Product Code Brand MWCO Quantity Price Quantity & Availability  
12644863
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T91
3 kDa 12/Pk.
£210.00
Pack of 12
In Stock
to see stock.
Add to basket
 
12684863
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T41
100 kDa 12/Pk.
£212.00
Pack of 12
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
 
12643465
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T11
5 kDa 12/Pk.
£216.00
Pack of 12
In Stock
to see stock.
Add to basket
 
12664863
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T31
50 kDa 12/Pk.
£216.00
Pack of 12
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
 
16696752
View Documents
Sartorius
VS15T01
10 kDa 12/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12663465
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T01
10 kDa 12/Pk.
£365.00
Pack of 12
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
12683465
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T21
30 kDa 12/Pk.
£233.00
Pack of 12
In Stock
to see stock.
Edge
Add to basket
 
12673465
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T02
10 kDa 48/Pk.
£793.00
Pack of 48
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
 
16616762
View Documents
Sartorius
VS15T02
10 kDa 48/Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12653465
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T12
5 kDa 48/Pk.
£793.00
Pack of 48
In Stock
to see stock.
Add to basket
 
12693465
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T22
30 kDa 48/Pk.
£793.00
Pack of 48
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
 
12694863
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T42
100 kDa 48/Pk.
£773.00
Pack of 48
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
 
12674863
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T32
50 kDa 48/Pk.
£818.00
Pack of 48
Estimated Shipment: 03-07-2025
to see stock.
Add to basket
 
12654863
BIOPHARMA
View Documents Promotion Details
Sartorius
VS15T92
3 kDa 48/Pk.
£773.00
Pack of 48
In Stock
to see stock.
Add to basket
 
Description

Description

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.

Twin vertical membranes and a sleek internal design for ultra-fast concentration
  • In a single spin, 15 ml solutions can be concentrated up to 150x in 10-30 minutes
  • Large, high-flux PES membranes feature ultra-low protein binding and low extractables
  • High, constant filtration pressure allows optimal crossflow to minimize membrane fouling
Designed for maximum convenience, safety and sample recovery
  • Angular, pipette-friendly pocket facilitates easy retentate recovery of greater than 95%
  • Eliminates the risk of sample filtration to dryness
  • Stable, inert polystyrene housing with high chemical compatibility (pH 1-14)
  • UV joining technology for a smooth transition between membrane and housing
  • Volume graduation for easy monitoring of concentrate volume and 100μl dead-stop volume
Universal compatibility for rotors that accept 50mL conical bottom tubes
  • Handles sample volumes up to 15mL in swing bucket rotors (11mL in fixed angle rotors)
  • Designed for rotors that accommodate Falcon 50mL conical tubes:
    • Beckman Allegra 25R with TS-5.1-500 swing-out rotor, BUC 5 buckets, adaptors
    • Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
    • Multifuge 3 S-R with 75006445 rotor, 75006441 buckets, adaptors
Specifications

Specifications

Single-use
Centrifuge
15 mL
7.2 cm2
Polystyrene
Centrifugal Concentrator
11.8 cm
Ultrafiltration
PES
Non-sterile
Concentration, desalting or buffer exchange applications
4,000 x g
11 mL
27 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Vivaspin™ Turbo 15 PES Centrifugal Concentrators

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.