Learn More
Sartorius Vivaspin™ Turbo 15 PES Centrifugal Concentrators

Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.
Brand: Sartorius VS15T11
140.99 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Combines high-flux, twin membranes with a sleek internal design for superior performance. Sartorius Vivaspin™ Turbo 15 Centrifugal Concentrators deliver ultra-fast concentration and virtually complete concentrate recovery. Available in a range of MWCO in 12-packs or convenient 48-pack cases.
Twin vertical membranes and a sleek internal design for ultra-fast concentration- In a single spin, 15 ml solutions can be concentrated up to 150x in 10-30 minutes
- Large, high-flux PES membranes feature ultra-low protein binding and low extractables
- High, constant filtration pressure allows optimal crossflow to minimize membrane fouling
- Angular, pipette-friendly pocket facilitates easy retentate recovery of greater than 95%
- Eliminates the risk of sample filtration to dryness
- Stable, inert polystyrene housing with high chemical compatibility (pH 1-14)
- UV joining technology for a smooth transition between membrane and housing
- Volume graduation for easy monitoring of concentrate volume and 100μl dead-stop volume
- Handles sample volumes up to 15mL in swing bucket rotors (11mL in fixed angle rotors)
- Designed for rotors that accommodate Falcon 50mL conical tubes:
- Beckman Allegra 25R with TS-5.1-500 swing-out rotor, BUC 5 buckets, adaptors
- Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors
- Multifuge 3 S-R with 75006445 rotor, 75006441 buckets, adaptors

Specifications
Single-use | |
Centrifuge | |
5 kDa | |
Non-sterile | |
Concentration, desalting or buffer exchange applications | |
4,000 x g | |
Centrifugal Concentrator | |
11.8 cm |
Ultrafiltration | |
PES | |
15 mL | |
7.2 cm2 | |
Polystyrene | |
12/Pk. | |
11 mL | |
27 mm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.